Flessen, brede hals, vierkant met schroefdraad, serie 310
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
310-70538 310-70539 310-70540
215-3312EA 2 EUR
215-3312 215-3313 215-3314
Flessen, brede hals, vierkant met schroefdraad, serie 310
Flessen Kunststof flessen
Natuur HDPE, zonder schroefdoppen.
  • Easy filling
  • Secure grip
  • Rectangular labelling field
  • Leakproof
  • Suitable for food
  • Halal certified
The wide neck allows for easy filling, emptying, and cleaning, Ribbed sides for secure grip.
Bestelinformatie:
Bottles are supplied without caps. Closures must be ordered separately.