Macron Fine Chemicals™ CHEM-SOLV®, Laboratory Glassware Cleaner
Leverancier: MACRON AVANTOR BRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2157-04EA 139.35 EUR
2157-04
Macron Fine Chemicals™ CHEM-SOLV®, Laboratory Glassware Cleaner
Reinigingsmiddelen Detergenten en oppervlaktereinigers
CHEM-SOLV® is a non chromic acid cleaner suitable for removing silicone greases, dried blood, and other organic residues from laboratory glass and plasticware. It is environmentally friendly and safer alternative for chromic and sulphuric acid solution.
  • Free of interfering ions
  • Ideal for cleaning labware used in sensitive analytical procedures and organic analysis
  • Cleans glassware without scrubbing