Silver(I) oxide ≥99%, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: di-Silver oxide, Disilver oxide, Silver oxide

21584.134EA 127 EUR
21584.134
Silver(I) oxide ≥99%, TECHNICAL
Silver(I) oxide
Formule: Ag₂O
Smeltpunt: 800…830 °C
Dichtheid: 7,14 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003404
CAS nummer: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1,II
Merck Index: 12,08665

Order Now

Specification Test Results

Assay Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR