Instrumentenschalen
Leverancier: P.S. KUNSTSTOFWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2035150010 2035420010 2049450010 2033910010 2034810010 2034030010
216-1676EA 0 EUR
216-1676 216-1677 216-1678 216-1679 216-1680 PSKA2034030010
Instrumentenschalen
Reservoirs/Schalen/Potten Instrumentenschalen
Melamine, white, polished.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Robust, smooth surfaces for easy cleaning
Low profile.