Tanks, cilindrisch, met deksel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11200-0005 11200-0007 11200-0010 11200-0015 11200-0030 11200-0055 11200-0100
216-1900EA 297 EUR
216-1900 216-1901 216-1902 216-1903 216-1904 216-1905 216-1906
Tanks, cilindrisch, met deksel, Nalgene®
Reservoirs Cilindrische tanks
These cylindrical polypropylene tanks with cover can resist stress cracking and increase tolerance to many organic chemicals.
  • An economic alternative to stainless-steel tanks
Bestelinformatie: Factory-installed spigots and tank liners are sold separately.