Mediaflessen, vierkant, steriel, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
382019-0030 382019-0125 382019-0060 382019-0250 382019-0500 382019-1000 382019-2000
216-2263EA 604 EUR
216-2263 216-2265 216-2264 216-2266 216-2267 216-2268 216-2269
Mediaflessen, vierkant, steriel, met schroefdop, Nalgene®
Flessen Mediumflessen
Certified clean container is made of PETG and comes with a sterile, natural HDPE closure with a tamper-resistant seal.
  • Non-cytotoxic, non-pyrogenic, non-hemolytic; gamma irradiated to a 10-⁶ Sterility Assurance Level
  • Leakproof bottle is free of animal derived components (ADCF) and lot certified for low particulate limits
  • Sterile
Manufactured in a certified ISO 14644-1 Class 7 cleanroom and packaged in cleanroom grade heat-sealed bags to assure product integrity. Suitable for pharmaceutical and biotechnology applications that require processing andMore Product Information