Boxes with Snap-On Lid, Multipurpose
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6210-1001 6210-2000 6210-0250 6210-0500 6250-1001 6250-1500 6250-2300 6250-3300 6250-2000 6250-1000 6250-0500
BURK6210-1001EA 5.6 EUR
BURK6210-1001 216-2984 BURK6210-0250 BURK6210-0500 BURK6250-1001 BURK6250-1500 BURK6250-2300 BURK6250-3300 BURK6250-2000 BURK6250-1000 BURK6250-0500
Boxes with Snap-On Lid, Multipurpose
Dozen Opbergdozen
Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (−40 °C up to briefly +95 °C).
PE, translucent.
  • Can be deep frozen
  • No crevices
  • Suitable for use with foodstuffs
Order Now
Specificaties
Environmentally Preferable