VWR® 32-Place PCR Racks

Leverancier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

216-2988EA 47.9 EUR
216-2988
VWR® 32-Place PCR Racks
Rekken Tube Racks
Designed for pre- and post-PCR sampling, streamline your PCR workflow with the 32-Place PCR rack. This versatile rack accommodates 0,2 ml tubes both individually and in strips of 8, as well as 32-place assay plates (8×4).

  • Small and compact design is stackable, saving valuable space in the freezer (Low temperature range: −80 °C)
  • Right sized when processing smaller batches
  • Lid hinge design allows lid to sit at 180 degrees, clear of sample processing, or be removed entirely
  • Transparent lid protects samples and allows quick assessment of contents
  • Embossed
More Product Information
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR