Potjes met schroefdop
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
916 914 910 912 913 911
216-3112EA 102 EUR
216-3112 216-3110 216-3102 216-3106 216-3108 216-3104
Potjes met schroefdop
Monsternamepotjes Containers voor algemeen gebruik
To store and transport samples securely.
LDPE, translucent.
  • Good chemical resistance, good temperature resistance
  • Strong walls, slightly conical shape
  • Particularly tight seal through screw cap with cone gasket