Compacte jerrycans met schroefdop, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0435-2050 0435-1050 0435-3000 0435-0001 0435-0010 0435-1100 0435-2100 0435-3010
216-5120EA 39 EUR
216-5120 216-5384 216-0898 216-0897 216-2192 216-2190 216-2191 216-2193
Compacte jerrycans met schroefdop, LaboPlast®
Flessen Jerrycans
Doorschijnend, PP.
  • With or without threaded connector ³/₄"
  • Tamper-proof closure
  • Canister without stopcock autoclavable
  • Moulded, indestructible litre graduation
This compact jerrycan is ideal as a space-saving canister (only 65 or 125 mm wide). Different liquids can be stored and decanted in a small space. It can also be stored lying on its side e.g. in a refrigerator. It is very stable due to ribbed structure and thick sides.
Compact jerrycan and Stopcock set
The Compact jerrycan and StopCock set consists of a Compact jerrycan with 5 or 10 l glandsMore Product Information
Order Now
Specificaties
Environmentally Preferable