Vaten met brede hals
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
359-90800 359-90867 359-91093
216-5652EA 64.8 EUR
216-5652 216-5541 216-5542
Vaten met brede hals
Vaten, vaten, trommels
HDPE. With lid and clamping ring.
  • Lid
  • Clamping ring
  • Suitable for food
  • All sizes with UN approval for solids
  • Halal certified
Barrels meet the GGVS/GGVE (rail/road/sea) regulations for the transport of solid substances with a density of at least 1,5 g/ccm. Supplied complete with PE lid and galvanized clamping ring. 30 and 60 L with handles.