Buckets, Round, LDPE
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0845
216-7339EA 151 EUR
216-7339
Buckets, Round, LDPE
Emmers
Round buckets, made of LDPE, with graduation, plastic coated metal handle and spout. Ideal for industrial applications.
  • Heavy, robust design
  • Good chemical resistance
  • Temperature resistance to 80 °C