Cetrimonium bromide, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

219374-100
219374-100EA 86.6 EUR
219374-100
Cetrimonium bromide, Millipore®
Cetrimonium bromide
Formule: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Smeltpunt: 237…243 °C
Vlampunt: 244 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011772
CAS nummer: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR