Decontamination bags, activated charcoal
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22007
22007.EA 140 EUR
22007.
Decontamination bags, activated charcoal
Reagentia voor elektroforese Reagentia voor kleurverwijdering
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
  • Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
  • Use to clean up any dyes from a solution to allow drain disposal
  • No-mess, convenient bag
  • Reusable bag capacity of at least 10 mg EtBr