Tyramide amplification buffer plus
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22029
22029.EA 725 EUR
22029.
Tyramide amplification buffer plus
Koppelings- en labelingssystemen voor eiwitten
An optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections.
  • For cell and tissue staining with tyramide signal amplification
  • Significantly improved brightness, specificity, and sensitivity
  • Compatible with CF® Dye and other tyramides
  • 250 or 1000 reaction sizes
  • 1× Tyramide amplification buffer plus
  • 30% Hydrogen peroxide
Tyramide amplification buffer plus is an optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections. It provides significantly improved brightness, specificity, and sensitivity compared to Biotium's original ready-to-use tyramide amplification buffer.
Tyramide amplification buffer plus was developed as a new and improved version of Biotium's original ready-to-use tyramide amplification buffer. It is also a component in all of Biotium's tyramide amplification kits.