Sodium bromide ≥99%, Reagent Grade

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

220345-12KG
220345-12KGEA 631 EUR
220345-12KG
Sodium bromide ≥99%, Reagent Grade
Sodium bromide

  • ≥95% for general laboratory use.

Formule: NaBr
MW: 102.89 g/mol
Kookpunt: 1.393 °C (1.013 hPa)
Smeltpunt: 750 °C
Dichtheid: 3.2 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003475
CAS nummer: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Assay of Br 76.8 - 78.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR