Trichloorazijnzuur ≥99% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

22156.18 22156.30 22156.A1
22156.18EA 51.4 EUR
22156.18 22156.30 22156.A1
Trichloorazijnzuur ≥99% ACS
Trichloorazijnzuur

Purity: 99% CAS No.: 76-03-9 Molecular Formula: CCl3CO2H Formula Weight: 163.39 Physical Form: Crystalline Density: 1.629 pH: 1 Melting Point: 54-58° Boiling Point: 196° Beilstein Reference: 970119 Merck Reference: 14,9627 UN No.: 1839 MDL No.: MFCD00004177


Protein precipitation reagent

Formule: Cl₃CCOOH
Kookpunt: 196 °C
Smeltpunt: 54…58 °C
Dichtheid: 1,629
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004177
CAS nummer: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8,II
Merck Index: 14,9627
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR