Magnesium trifluoromethanesulphonate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

22340.09 22340.18
22340.09EA 60.3 EUR
22340.09 22340.18
Magnesium trifluoromethanesulphonate
Magnesium trifluoromethanesulphonate

CAS No.: 60871-83-2 Molecular Formula: C2F6MgO6S2 Formula Weight: 322.43 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 310° Beilstein Reference: 705578 UN No.: 1759 MDL No.: MFCD00042601

Formule: C₂F₆MgO₆S₂
Smeltpunt: >310 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042601
CAS nummer: 60871-83-2
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR