Slangen, polyurethaan, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8030-0060 8030-0120
NALG8030-0060EA 91.4 EUR
NALG8030-0060 224-2031
Slangen, polyurethaan, Nalgene®
Slangen Transfer Tubing
Polyurethane, translucent.
  • Contains no plasticizers and only low levels of extractables for high-purity applications
  • Superior chemical resistance to fuels, oils, and some solvents (not recommended for aqueous solutions)
  • Resistant to atmospheric ozone, aliphatic hydrocarbons and petroleum products for longer life and versitility
  • This transparent, non-autoclavable tubing is marked every 12" for cutting and can be gas sterilized
  • Ideal for peristaltic pump applications
Polyurethane tubing, which exhibits excellent abrasion and flex-fatigue resistance, asMore Product Information