Calcium aluminate ≥99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

22646.06 22646.14 22646.22
22646.06EA 71.8 EUR
22646.06 22646.14 22646.22
Calcium aluminate ≥99% (metals basis)
Calcium aluminate

Purity: 99% CAS No.: 12042-68-1 Molecular Formula: CaAl2O4 Formula Weight: 158.02 Physical Form: -200 Mesh Powder Density: 2.981 Melting Point: 1600° MDL No.: MFCD00049722

Formule: Al₂CaO₄
Smeltpunt: 1600 °C
Dichtheid: 2,981
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049722
CAS nummer: 12042-68-1
EINECS: 234-931-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR