Chloramine-T (sodium salt) trihydrate ≥98% (by iodometry), GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

22685.265EA 56.5 EUR
22685.265 22685.360 22685.460
Chloramine-T (sodium salt) trihydrate ≥98% (by iodometry), GPR RECTAPUR®
Chloramine-T (sodium salt) trihydrate
Formule: CH₃C₆H₄SO₂NClNa·3H₂O
Smeltpunt: 167…170 °C
Dichtheid: 1,43 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149066
CAS nummer: 7080-50-4
EINECS: 204-854-7
UN: 3263
ADR: 8,III
Merck Index: 13,02085

Order Now

Specification Test Results

Assay (iodometric) ≥ 98 %
Identification (IR) Passes test
pH (25°C; 5 %) 8.0 - 11.5
Alcohol-insoluble matter ≤ 1.5 %
Heavy metals (as Pb) ≤ 100 ppm
Fe (Iron) ≤ 100 ppm
Residue insoluble in anhydrous ethanol. Max. 2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR