Chloramine-T (sodium salt) trihydrate 98,0-103, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt

22686.230EA 33.4 EUR
22686.230
Chloramine-T (sodium salt) trihydrate 98,0-103, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Chloramine-T (sodium salt) trihydrate
Formule: CH₃C₆H₄SO₂NClNa·3H₂O
MW: 281.69 g/mol
Vlampunt: 192
MDL Nummer: MFCD00149066
CAS nummer: 7080-50-4
UN: 3263
ADR: 8 III
Merck Index: 13,02085

Order Now

Specification Test Results

Assay 98.0 - 103.0 %
Identification A Passes test Reag.Ph.Eur.
Identification B Passes test Reag.Ph.Eur.
Identification C Passes test Reag.Ph.Eur.
Identification D Passes test Reag.Ph.Eur.
Identification E Passes test Reag.Ph.Eur.
Appearance of solution Passes test Reag.Ph.Eur.
Clarity of aqueous solution Passes test ACS
Ortho compounds Passes test
Suitability for determination of bromide Passes test ACS
pH (20°C; 5 %) 8.0 - 10.0
pH (25°C; 5 %) 8.0 - 10.0
Residue insoluble in anhydrous ethanol Max. 1.5 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR