Slangen, silicone, HelixMark®
Leverancier: Freudenberg Medical Europe GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60-011-01 60-011-02 60-011-23 60-011-03 60-011-08 60-011-09 60-011-10 60-011-11 60-011-21 60-011-22 60-011-24 60-011-25 60-011-26 60-011-27 60-011-28 60-011-29 60-011-31 60-011-32 60-011-33 60-011-34 60-111-21 60-111-24 60-111-25 60-111-29 60-111-30 60-111-31 60-111-32 60-111-33 60-011-04 60-011-05 60-011-06 60-011-07 60-011-30 60-111-22 60-011-05-70MM 60-011-06-70MM 60-011-07-70MM 60-011-35
228-0654EA 76.7 EUR
228-0654 228-0670 228-0659 228-1065 228-1066 228-1067 228-1068 228-1069 228-1070 228-1071 228-1072 228-1073 228-1074 228-1075 228-1076 228-1077 228-1078 228-1079 228-1080 228-1081 228-1082 228-1084 228-1085 228-1086 228-1087 228-1088 228-1089 228-1090 228-0655 228-0656 228-0657 228-0658 228-0660 228-1083 HELI60-011-05-70MM HELI60-011-06-70MM HELI60-011-07-70MM HELI60-011-35
Slangen, silicone, HelixMark®
Slangen Transfer Tubing
Silicone tubing, translucent, platinum cross-linked, with very tight tolerances. All HelixMark products are manufactured in clean rooms in compliance with the requirements of the medtech and pharmaceutical industry.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Supplied with material certificate in every packaging unit
  • Seamless batch traceability
HelixMark standard silicone tubing meets the needs of a broad spectrum of medical andMore Product Information
Order Now
Specificaties
Environmentally Preferable