HelixMark® Peristaltic Pump Tubing
Leverancier: Freudenberg Medical Europe GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
60-825-163 60-825-245 60-825-110 60-825-130 60-825-133 60-825-136 60-825-139 60-825-141 60-825-160 60-825-166 60-825-191 60-825-194 60-825-200 60-825-230 60-825-243 60-825-248 60-825-27 60-825-288 60-825-310 60-825-37 60-825-420 60-825-45 60-825-51 60-825-63 60-825-70 60-825-82 60-825-93 60-825-104 60-825-197 60-825-428 60-825-108 60-825-112 60-825-15 60-825-23 60-825-262 60-825-270 60-825-43 60-825-79 49303830 49321568 60-825-79.250M 61-395-02 61-395-01 49304287 60-825-231 60-825-310.15M
228-0668EA 182 EUR
228-0668 228-0653 228-1111 228-1112 228-1113 228-1114 228-1115 228-1116 228-1118 228-1119 228-1120 228-1121 228-1122 228-1124 228-1125 228-1126 228-1128 228-1129 228-1130 228-1131 228-1132 228-1134 228-1135 228-1136 228-1137 228-1139 228-1140 228-0688 228-0689 228-0690 228-0666 228-0667 228-1117 228-1123 228-1127 228-0669 228-1133 228-1138 228-1363 228-1569 224-0177 228-1390 HELI61-395-01 HELI49304287 HELI60-825-231 HELI60-825-310.15M
HelixMark® Peristaltic Pump Tubing
Slangen Transfer Tubing
HelixMark peristaltic pump tubing is designed and manufactured for use with peristaltic pumps and other applications with higher effective forces. A special production process designed to meet these requirements ensures that the products provide greater stability and capacity to withstand stresses. The range offers sizes suitable for use with all commonly used pump types. The tubing is individually packed in 7.5 m coils.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene
More Product Information