Cobalt(II) chloride hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent nickel-free

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

22896.184EA 198 EUR
22896.184
Cobalt(II) chloride hexahydrate 98.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent nickel-free
Cobalt(II) chloride hexahydrate
Formule: CoCl₂·6H₂O
MW: 237.93 g/mol
Kookpunt: 110
Smeltpunt: 56
Dichtheid: 1.92 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149652
CAS nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Insolubility in water Max. 100 ppm
NO3 (Nitrate) Max. 100 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 20 ppm
Mn (Manganese) Max. 10 ppm
Na (Sodium) Max. 100 ppm
Ni (Nickel) Max. 50 ppm
Pb (Lead) Max. 5 ppm
Zn (Zinc) Max. 20 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR