Methanol 99,8, ultrapure HPLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

22909.K2
22909.K2EA 69.2 EUR
22909.K2
Methanol 99,8, ultrapure HPLC
Methanol

Purity: 99.8+% CAS No.: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3290 Odour: Mild Melting Point: -98° Boiling Point: 64.7° Beilstein Reference: 1098229 Merck Reference: 14,5957 UN No.: 1230 MDL No.: MFCD00004595

Formule: CH₃OH
MW: 32.04 g/mol
Kookpunt: 64.7 °C
Smeltpunt: –98 °C
Dichtheid: 0.791
Vlampunt: 11 °C (52 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
Bereik: 01-2119433307-44
Merck Index: 14,5957
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR