Copper(II) chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

23093.233EA 64 EUR
23093.233 23093.290
Copper(II) chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Copper(II) chloride dihydrate
Formule: CuCl₂·2H₂O
Smeltpunt: 100 °C
Dichtheid: 2,54 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149674
CAS nummer: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (20°C; 5 %) 3.0 - 3.8
Insolubility in water Max. 100 ppm
Total N (Nitrogen) Max. 30 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 1 ppm
Fe (Iron) Max. 30 ppm
Ni (Nickel) Max. 10 ppm
Pb (Lead) Max. 40 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR