Pharmacist's spatulas
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
661.300 661.250 661.200 661.150 661.130 661.100 661.075 061.150 061.130 061.100 061.075
231-0312EA 32.3 EUR
231-0312 231-0300 231-0299 231-0554 231-0553 231-0552 231-0551 231-0550 231-0549 231-0548 231-0547
Pharmacist's spatulas
Spatels
Stainless steel blade, wooden handle.
  • Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
  • 75 to 150 mm: conical blade
Order Now
Specificaties
Environmentally Preferable