Apothekerspatels
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
851.130 851.100 851.075 861.075 861.100 851.150 861.150 861.130
231-0557EA 59.3 EUR
231-0557 231-0556 231-0555 231-0318 231-0319 231-0558 231-0321 231-0320
Apothekerspatels
Spatels
Chromium steel, hardened, extremely flexible blade.
  • Excellent thermal resistance
  • Good chemical resistance
  • Quick and easy maintenance
  • Available with wooden handle or with dishwasher-safe plastic handle