Powder spatulas, disposable, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8095A-150 8095A-150S 8095A-250 8095A-250S 8095R-150S 8095R-150 8095R-250 8095R-250S
231-3506EA 161 EUR
231-3506 231-3507 231-3508 231-3509 SAMP8095R-150S SAMP8095R-150 SAMP8095R-250 SAMP8095R-250S
Powder spatulas, disposable, SteriWare®
Spatels
PS. These disposable spatulas feature a rounded handle and a robust blade.
  • Manufactured and packed in a controlled atmosphere (class 100000 cleanroom)
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Approved for use in ATEX Zones 0, 1 and 2 (contact Sampling Systems for full details)
  • Disposable: No cleaning is required before or after sampling
  • Available as gamma irradiated sterile (SAL 10-6)
Suitable for sterile sampling and dispensing quantities of powders in all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.).