Cobalt(II) bromide, watervrij 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

23149.18 23149.30
23149.18EA 88.2 EUR
23149.18 23149.30
Cobalt(II) bromide, watervrij 97%
Cobalt(II) bromide

Purity: 97% CAS No.: 7789-43-7 Molecular Formula: CoBr2 Formula Weight: 218.75 Physical Form: Crystalline Density: 4.909 Appearance: Green Melting Point: 678°(in N2) Solubility: Soluble in water, methanol, ethanol, acetone, methyl acetate Merck Reference: 14,2435 MDL No.: MFCD00016017

Formule: Br₂Co
Smeltpunt: 678 °C(in N{2})
Dichtheid: 4,909
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016017
CAS nummer: 7789-43-7
EINECS: 232-166-7
Merck Index: 14,2435

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR