4-Hydroxy-4-methyl-2-pentanon ≥98.0%, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

23427.290EA 93.4 EUR
23427.290 23427.320 23427.460
4-Hydroxy-4-methyl-2-pentanon ≥98.0%, GPR RECTAPUR®
4-Hydroxy-4-methyl-2-pentanon
Formule: (CH₃)₂C(OH)CH₂COCH₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004471
CAS nummer: 123-42-2
EINECS: 204-626-7
UN: 1148
Bereik: 01-2119473975-21

Order Now

Specification Test Results

Assay ≥ 98,0 %
Identification (IR) Passes test Passes test
Colouration ≤ 20 APHA ≤ 20 APHA
Density (20/4) 0,937 - 0,943
Refractive index (20°C) 1,422 - 1,425
Evaporation residue ≤ 0,02 %
Water ≤ 0,1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR