4-(Dimethylamino)benzaldehyde ≥99%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

23439.158EA 114 EUR
23439.158 23439.238
4-(Dimethylamino)benzaldehyde ≥99%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
4-(Dimethylamino)benzaldehyde
Formule: (CH₃)₂NC₆H₄CHO
Kookpunt: 266 °C (1013 hPa)
Smeltpunt: 72 °C
Dichtheid: 1,0254 g/cm³ (99.9 °C)
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003381
CAS nummer: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Bases of indole type Passes test
IR Spectrum Passes test
Organic impurities Passes test
Solution in acetic acid Passes test
Melting point 72 - 74 °C
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.1 %
Insolubility in alcohol Max. 100 ppm
Cl (Chloride) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 5 ppm
Conforms to Reag. Ph.Eur. Passes test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR