Waterstofperoxide 3% Stabilised, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


Stabilisatie: 0.04 % Phosphat

23614.291EA 59.4 EUR
23614.291
Waterstofperoxide 3% Stabilised, GPR RECTAPUR®
Waterstofperoxide
Formule: H₂O₂
MW: 34.01 g/mol
Kookpunt: 150.2
Smeltpunt: -0.41
Dichtheid: 1.01 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011333
CAS nummer: 7722-84-1
EINECS: 231-765-0
Merck Index: 14,04798

Order Now

Specification Test Results

Assay 2.5 - 3.5 %
Evaporation residue Max. 0.05 %
Heavy metals (as Pb) Max. 5 ppm
Cl (Chloride) Max. 10 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR