Tin(II) chloride dihydrate ≥96%, GPR RECTAPUR® for mirror makers

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Stannous chloride dihydrate

23743.230EA 62.9 EUR
23743.230 23743.296
Tin(II) chloride dihydrate ≥96%, GPR RECTAPUR® for mirror makers
Tin(II) chloride dihydrate
Formule: SnCl₂·2H₂O
Kookpunt: 623 °C (1013 hPa)
Smeltpunt: 246 °C
Dichtheid: 2,71 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149863
CAS nummer: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay Min. 96 %
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 100 ppm
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR