Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

24103-2.5L 24103-1L 24103-30KG 24103-5L 24103-4X5L 24103-6X1L 24103-4X2.5L
24103-2.5LEA 0 EUR
24103-2.5L 24103-1L 24103-30KG 24103-5L 24103-4X5L 24103-6X1L 24103-4X2.5L
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Ethanol

Ethanol (Untaxed), sales not in Germany, for license holders only

Formule: C₂H₅OH
Kookpunt: 78…79 °C (1,013 hPa)
Smeltpunt: –115 °C
Dichtheid: 0,790…0.791 g/cm3 (20 °C)
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003568
CAS nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
Bereik: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (GC) Min. 99.8 Vol.%
Density (D 20/20) 0.790 - 0.791
Non-volatile matter Max. 0.01 %
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0005 %
Aldehydes (as CH₃CHO) Max. 0.005 %
Methanol (GC) Max. 0.1 %
S-compounds (as S) max. 0.2 ppm
Explosion Limit 19 %, 60 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR