n-Hexaan 95, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Hexyl hydride, Hexanen, Hexane

24611.297EA 24.3 EUR
24611.297 24611.366 24611.468 24611.550
n-Hexaan 95, TECHNICAL
n-Hexaan
Formule: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Kookpunt: 69
Smeltpunt: –94,3 °C
Dichtheid: 0.659 g/cm³ (20 °C)
Vlampunt: –22 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009520
CAS nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Bereik: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 95 %
Boiling point 68 - 70 °C
Benzene Max. 100 ppm
Evaporation residue Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR