Lactose monohydrate, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Milk sugar, Lactobiose

24945.291EA 40 EUR
24945.291 24945.360
Lactose monohydrate, GPR RECTAPUR®
Lactose monohydrate
Formule: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Smeltpunt: 223
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 10039-26-6
EINECS: 200-559-2

Order Now

Specification Test Results

Spec. opt. rotation (calc. on anhydrous) 54.4 - 55.9 °
Ignition residue (SO4) Max. 0.1 %
Loss on drying (120°C) 4.0 - 5.5 %
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR