Anti-CD11c Mouse Monoclonal Antibody (APC-Cyanine7) [clone: 3.9]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25-0116-T025 25-0116-T100
25-0116-T025EA 158 EUR
25-0116-T025 25-0116-T100
Anti-CD11c Mouse Monoclonal Antibody (APC-Cyanine7) [clone: 3.9]
Antilichamen
The 3.9 antibody reacts with human CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18)More Product Information