Corning® Trypsin

Leverancier: Corning
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

25-051-CI
25-051-CIEA 77.87 EUR
25-051-CI
Corning® Trypsin
Enzymen

0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.


  • Porcine parvovirus tested
  • 6×100 ml


Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances thh efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and contains sodium bicarbonate.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR