Corning® Lymphocyte Separation Medium, Corning
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25-072-CV 25-072-CI
25-072-CVEA 151.65 EUR
25-072-CV MDTC25-072-CI
Corning® Lymphocyte Separation Medium, Corning
Media voor celscheiding
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.