Magnesium sulphate, anhydrous ≥99%, powder, BAKER ANALYZED®, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: MgSO4, Magnesium sulphate anhydrous

2506-01EA 178.11 EUR
2506-01 2506-05 2506-R
Magnesium sulphate, anhydrous ≥99%, powder, BAKER ANALYZED®, J.T.Baker®
Magnesium sulphate

The USP/NF Offical Compendia of Standards contains a section titled, "Reagents, Indicators, and Solutions" that lists the quality requirements of chemicals to be used in compendia testing. PharmaTest reagents are tested according to these requirements, relieving you of the need to do additional testing on your reagents. Products may be reagent or other grade, but are labeled as "Meets reagent specifications for testing USP/NF monographs." Test solutions are labeled as PharmaTest.

Formule: MgSO₄
MW: 120.37 g/mol
Smeltpunt: 1127
Dichtheid: 2.65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011110
CAS nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥99.0%
Assay (MgSO₄) (ignited basis) ≥99.0 %
Loss on Ignition ≤2.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.004 %
Ammonium (NH₄) ≤0.004 %
Calcium (Ca) ≤0.06 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Manganese (Mn) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR