Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Stannous chloride dihydrate

2512.0250EA 86.6 EUR
2512.0250
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Tin(II) chloride dihydrate
Formule: SnCl₂·2H₂O
MW: 225.65 g/mol
Kookpunt: 623
Smeltpunt: 246
Dichtheid: 2.71 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149863
CAS nummer: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay Min. 98 %
Insolubility in diluted HCl Passes test
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 20 ppm
As (Arsenic) Max. 1 ppm
Ca (Calcium) Max. 0,005%
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 20 ppm
Hg (Mercury) Max. 0.05 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 100 ppm
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR