Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

252531000 252538000
252531000.EA 75 EUR
252531000. 252538000.
Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Sodium-tert-pentyloxide
Formule: C₅H₁₁NaO
Dichtheid: 0,91 g/cm³
Vlampunt: –11 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064245
CAS nummer: 14593-46-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration with HClO4 ≥30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR