Paraffin, Solidification point: 52-54°C, pellets Ph. Eur.

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Wax Paraffin, Paraffin wax

26157.291EA 49.1 EUR
26157.291 26157.460
Paraffin, Solidification point: 52-54°C, pellets Ph. Eur.
Paraffin
Smeltpunt: 52…54 °C
Dichtheid: 0,9 g/cm³ (20 °C)
CAS nummer: 64742-51-4
EINECS: 232-315-6

Order Now

Specification Test Results

Appearance White mass
Identification A Passes test
Melting point 50 - 61 °C
Acidity or alkalinity Passes test
Polycyclic aromatic hydrocarbons Passes test
SO4 (Sulphate) Max. 150 ppm
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR