DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus), Calbiochem®, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

2618-1 2618-250
2618-1EA 412 EUR
2618-1 2618-250
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus), Calbiochem®, Millipore®
DNA sodium salt (Deoxyribonucleic acid sodium salt)

It is a purified DNA from calf thymus and an essential component of chromosomes


Special Instructions: Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00130921
CAS nummer: 9007-49-2

Order Now

Specification Test Results

hyperchromicity ≥27%
S20w 15.4-17.4
A260/280 nm ≥ 1.8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR