Sodium deoxycholate, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate

264101-1 264101-100 264101-25
264101-1EA 1240 EUR
264101-1 264101-100 264101-25
Sodium deoxycholate, Millipore®
Sodium deoxycholate
Formule: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Smeltpunt: 357…365 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064139
CAS nummer: 302-95-4
EINECS: 206-132-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR