Kaliumhydroxide ≥85%, pellets, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

26669.266EA 26.6 EUR
26669.266 26669.290 26669.368 26669.460
Kaliumhydroxide ≥85%, pellets, GPR RECTAPUR®
Kaliumhydroxide
Formule: KOH
Kookpunt: 1320 °C (1013 hPa)
Smeltpunt: 360 °C
Dichtheid: 2,04 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
Bereik: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Assay Min. 85 %
Total N (Nitrogen) Max. 5 ppm
Cl (Chloride) Max. 50 ppm
CO3 (as K2CO3) Max. 2.0 %
PO4 (Phosphate) Max. 20 ppm
SiO4 (as SiO2) Max. 100 ppm
SO4 (Sulphate) Max. 50 ppm
Al (Aluminium) Max. 10 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR