Kaliumpermanganaat 99, GPR RECTAPUR®

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Permanganic acid potassium salt

26906.290EA 75.8 EUR
26906.290 26906.368
Kaliumpermanganaat 99, GPR RECTAPUR®
Kaliumpermanganaat
Formule: KMnO₄
MW: 158.03 g/mol
Smeltpunt: > 240 °C
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011364
CAS nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay Min. 99 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.02 %
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR