DNA (Deoxyribonucleic acid) (from salmon testes), sonicated

Leverancier: Cytiva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

27-4565-01
27-4565-01EA 514 EUR
27-4565-01
DNA (Deoxyribonucleic acid) (from salmon testes), sonicated
DNA (Deoxyribonucleic acid)

Genomic DNA sheared by sonication for use in hybridization experiments


  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • Suitable for blocking non-specific binding sites in hybridization experiments to reduce background


Sonicated DNA from salmon testes.

Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00130922
CAS nummer: 9007-49-2

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR